missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXYD5/Dysadherin Antibody (CL14039) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24586G
This item is not returnable.
View return policy
Description
FXYD5/Dysadherin Monoclonal antibody specifically detects FXYD5/Dysadherin in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| FXYD5/Dysadherin | |
| Monoclonal | |
| DyLight 488 | |
| DYSAD, dysadherin, FXYD domain containing ion transport regulator 5, FXYD domain-containing ion transport regulator 5, IWU1, KCT1, keratinocytes associated transmembrane protein 1, OIT2, PRO6241, RIC | |
| This antibody was developed using a recombinant protein derived from Q96DB9, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Signal Transduction | |
| 53827 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL14039 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction