missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXYD5/Dysadherin Antibody (Centrose patent anti-dysadherin) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28555JF585
This item is not returnable.
View return policy
Description
FXYD5/Dysadherin Monoclonal antibody specifically detects FXYD5/Dysadherin in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| FXYD5/Dysadherin | |
| Monoclonal | |
| Janelia Fluor 585 | |
| DYSAD, dysadherin, FXYD domain containing ion transport regulator 5, FXYD domain-containing ion transport regulator 5, IWU1, KCT1, keratinocytes associated transmembrane protein 1, OIT2, PRO6241, RIC | |
| Dysadherin | |
| 0.1 mL | |
| Signal Transduction | |
| 53827 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Centrose patent anti-dysadherin | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction