missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXYD3 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98625C
This item is not returnable.
View return policy
Description
FXYD3 Polyclonal antibody specifically detects FXYD3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FXYD3 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 5349 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| Chloride conductance inducer protein Mat-8, FXYD domain containing ion transport regulator 3, FXYD domain-containing ion transport regulator 3, Mammary tumor 8 kDa protein, MAT8MAT-8, Phospholemman-like, phospholemman-like protein, PLMLMGC111076 | |
| Produced in rabbits immunized with E. coli-derived Human FXYD3 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction