Learn More
FXYD1 Mouse, anti-Human, CoraLite Plus 488, Clone: 2C10G9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67150-100UL
Description
The FXYD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FXYD1. This antibody recognizes human, pig, mouse antigen. The FXYD1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
| FXYD1 | |
| 2C10G9 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Pig, Mouse | |
| 155 kDa | |
| IgG2a |
| Flow Cytometry (Intracellular Staining) | |
| 1000 μg/mL | |
| FXYD1, Phospholemman, PLM | |
| 100 μL | |
| 5348 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |