missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03963-100UL
This item is not returnable.
View return policy
Description
FXR2 Polyclonal specifically detects FXR2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| FXR2 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| FMR1L2, fragile X mental retardation syndrome-related protein 2, fragile X mental retardation, autosomal homolog 2, fragile X-mental retardation 1-like 2, FXR2P | |
| A synthetic peptide corresponding to a sequence within amino acids 550-650 of human FXR2 (NP_004851.2). | |
| 100 μL | |
| Cellular Markers, Chromatin Research, Neurodegeneration, Neuroscience | |
| 9513 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction