missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXR2 Antibody (1G2), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-259B
This item is not returnable.
View return policy
Description
FXR2 Monoclonal specifically detects FXR2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| FXR2 | |
| Monoclonal | |
| Biotin | |
| P51116 | |
| FXR2 | |
| Recombinant C-terminal fragment of human FXR2 (residues 414-658). [UniProt# P51116] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1G2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| FMR1L2, fragile X mental retardation syndrome-related protein 2, fragile X mental retardation, autosomal homolog 2, fragile X-mental retardation 1-like 2, FXR2P | |
| Mouse | |
| 80 kDa | |
| 0.1 mL | |
| Cellular Markers, Chromatin Research, Neurodegeneration, Neuroscience | |
| 9513 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction