missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FXR1 Recombinant Rabbit Monoclonal Antibody (23GB5465), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | FXR1 |
|---|---|
| Clone | 23GB5465 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088113
![]() |
Invitrogen™
MA552858 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089043
|
Invitrogen™
MA552859 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FXR1 Recombinant Monoclonal antibody specifically detects FXR1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| FXR1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P51114, Q5XI81, Q61584 | |
| 14359, 361927, 8087 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5465 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1110050J02Rik; 9530073J07Rik; AI851072; FMR1 autosomal homolog 1; fragile X mental retardation autosomal homolog variant p1K; fragile X mental retardation autosomal homolog variant p2K; fragile X mental retardation autosomal homolog variant p4K; fragile X mental retardation autosomal homolog variant p5FK; fragile X mental retardation gene 1, autosomal homolog; fragile X mental retardation gene, autosomal homolog; fragile X mental retardation syndrome-related protein 1; fragile X mental retardation, autosomal homolog 1; fragile X mental retardation-related protein 1; fragile-X-related protein 1; Fxr1; Fxr1h; Fxr1p; hFXR1p; hypothetical protein LOC536793; mFxr1p | |
| A synthesized peptide derived from human FXR1 (4-36AA). | |
| Primary | |
| -20°C | |
| Fxr1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title