missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FXR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596481
This item is not returnable.
View return policy
Description
Immunogen sequence: VTAIELDEDT GTFRIYGESA DAVKKARGFL EFVEDFIQVP RNLVGKVIGK NGKVIQEIVD KSGVVRVRIE GDNENKLPRE DGMVPFVFVG TKESIGNVQV L; Positive Samples: HeLa, HepG2, MCF7, Mouse brain, Mouse kidney, Mouse heart; Cellular Location: Cytoplasm.
FXR1 is an RNA binding protein that interacts with the functionally-similar proteins FMR1 and FXR2. It may regulate intracellular transport and local transltion of certain mRNAs. These proteins shuttle between the nucleus and cytoplasm and associate with polyribosomes, predominantly with the 60S ribosomal subunit. FXR1 is required for embryonic and postnatal development of muscle tissue. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| FXR1 | |
| Polyclonal | |
| Unconjugated | |
| Fxr1 | |
| 1110050J02Rik; 9530073J07Rik; AI851072; FMR1 autosomal homolog 1; fragile X mental retardation autosomal homolog variant p1K; fragile X mental retardation autosomal homolog variant p2K; fragile X mental retardation autosomal homolog variant p4K; fragile X mental retardation autosomal homolog variant p5FK; fragile X mental retardation gene 1, autosomal homolog; fragile X mental retardation gene, autosomal homolog; fragile X mental retardation syndrome-related protein 1; fragile X mental retardation, autosomal homolog 1; fragile X mental retardation-related protein 1; fragile-X-related protein 1; Fxr1; Fxr1h; Fxr1p; hFXR1p; hypothetical protein LOC536793; mFxr1p | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14359, 361927, 8087 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunofluorescence, Western Blot | |
| 2.07 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P51114, Q5XI81, Q61584 | |
| Fxr1 | |
| A synthetic peptide corresponding to a sequence within amino acids 250-350 of human FXR1 (NP_0050782). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction