missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXR/NR1H4 Antibody (OTI4F12), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71526PE
This item is not returnable.
View return policy
Description
FXR/NR1H4 Monoclonal antibody specifically detects FXR/NR1H4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| FXR/NR1H4 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI4F12 | |
| Western Blot, Immunohistochemistry | |
| bile acid receptor, farnesoid X receptor, Farnesoid X-activated receptor, Farnesol receptor HRR-1, FXRBAR, HRR-1, HRR1MGC163445, Nuclear receptor subfamily 1 group H member 4, nuclear receptor subfamily 1, group H, member 4, Retinoid X receptor-interacting protein 14, RIP14RXR-interacting protein 14 | |
| Human recombinant protein fragment corresponding to amino acids 21-297 of human NR1H4 (NP_005114) produced in E.coli. | |
| 0.1 mL | |
| Cancer, Chromatin Research, Lipid and Metabolism, Signal Transduction | |
| 9971 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction