missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXR/NR1H4 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB400-153AF350
This item is not returnable.
View return policy
Description
FXR/NR1H4 Polyclonal specifically detects FXR/NR1H4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| FXR/NR1H4 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| bile acid receptor, farnesoid X receptor, Farnesoid X-activated receptor, Farnesol receptor HRR-1, FXRBAR, HRR-1, HRR1MGC163445, Nuclear receptor subfamily 1 group H member 4, nuclear receptor subfamily 1, group H, member 4, Retinoid X receptor-interacting protein 14, RIP14RXR-interacting protein 14 | |
| Rabbit | |
| Affinity Purified | |
| Cancer, Chromatin Research, Lipid and Metabolism, Signal Transduction | |
| 9971.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| NR1H4 | |
| A synthetic peptide made to the C-terminus of the human FXR protein. [UniProtNo. Q96RI1] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction