Learn More
FXN Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14147-1-AP-20UL
This item is not returnable.
View return policy
Description
The FXN antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FXN. This antibody recognizes human, mouse, rat antigen. The FXN antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| FXN | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 2395 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 133 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |