missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FXC1 Polyclonal specifically detects FXC1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | FXC1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 646 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | fracture callus 1 (rat) homolog, fracture callus 1 homolog (rat), Fracture callus protein 1, FxC1, Tim10b, Tim9b, TIMM10Bmitochondrial import inner membrane translocase subunit Tim9 B, TIMM9B |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human FXC1 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?