missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXC1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-06236CL1
This item is not returnable.
View return policy
Description
FXC1 Polyclonal antibody specifically detects FXC1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FXC1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| fracture callus 1 (rat) homolog, fracture callus 1 homolog (rat), Fracture callus protein 1, FxC1, Tim10b, Tim9b, TIMM10Bmitochondrial import inner membrane translocase subunit Tim9 B, TIMM9B | |
| Produced in rabbits immunized with E. coli-derived Human FXC1 fragment. | |
| 0.1 mL | |
| Endocrinology, Signal Transduction | |
| 26515 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction