missing translation for 'onlineSavingsMsg'
Learn More

FX1, MedChemExpress

Product Code. 16440946
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16440946

Brand: Medchem Express HY102027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FX1 is a potent and specific BCL6 inhibitor, with an IC50 of around 35 μM.

FX1 is a potent and specific BCL6 inhibitor, with an IC50 of around 35 μM.

Specifications

Chemical Name or Material FX1
Physical Form Solid
CAS 1426138-42-2
Color Red
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C14H9ClN2O4S2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 10 mg/mL (27.11 mM; Need ultrasonic)
SMILES O=C(O)CCN(C/1=O)C(SC1=C2C(NC3=C/2C=C(Cl)C=C3)=O)=S
Molecular Weight (g/mol) 368.82
Formula Weight 368.82
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.