missing translation for 'onlineSavingsMsg'
Learn More

FX-11, MedChemExpress

Product Code. 16489547
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489547 5 mg 5mg
16477066 100 mg 100mg
16443057 50 mg 50mg
16437008 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16489547 Supplier Medchem Express Supplier No. HY16214
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress FX-11 is a potent, selective, reversible and competitive lactate dehydrogenase A (LDHA) specific inhibitor, with a Ki of 8 μM. FX-11 can effectively activate PKM2 (pyruvate kinase M2). FX-11 reduces ATP levels and induces oxidative stress, ROS production and cell death. FX-11 shows antitumor activity in lymphoma and pancreatic cancer xenografts.

FX-11 is a potent, selective, reversible and competitive lactate dehydrogenase A (LDHA) specific inhibitor, with a Ki of 8 μM. FX-11 can effectively activate PKM2 (pyruvate kinase M2). FX-11 reduces ATP levels and induces oxidative stress, ROS production and cell death. FX-11 shows antitumor activity in lymphoma and pancreatic cancer xenografts.

Specifications

Chemical Name or Material FX-11
CAS 213971-34-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H22O4
Quantity 5 mg
Synonym LDHA Inhibitor FX11
Solubility Information DMSO : 250 mg/mL (713.45 mM; Need ultrasonic)
SMILES O=C(C1=C2C=C(CC3=CC=CC=C3)C(C)=CC2=C(CCC)C(O)=C1O)O
Molecular Weight (g/mol) 350.41
Formula Weight 350.41
Percent Purity 98.44%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.