missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FUZ Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531699
This item is not returnable.
View return policy
Description
Recommended positive controls: HCT116. Predicted reactivity: Mouse (88%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FUZ is the human homolog of Drosophila fy protein and belongs to the fuzzy family. The fy gene of Drosophila encodes a novel four-pass transmembrane protein that plays two roles in the development of hairs on the Drosophila wing. The first is to specify the correct orientation of the hair by restricting its initiation to the distal vertex of the cell. The second role is to permit the development of just a single cell hair by maintaining the integrity of the F-actin and microtubule arrays that are required for hair development. In humans, the protein is involved in cytoskeleton function and cell polarity in epithelial cells. The human fuz gene is localized on chromosome 19q13.33.
Specifications
| FUZ | |
| Polyclonal | |
| Unconjugated | |
| FUZ | |
| 2600013E07Rik; b2b1273Clo; Fuz; fuzzy homolog; fuzzy homolog (Drosophila); fuzzy planar cell polarity protein; FY; NTD; Protein fuzzy homolog; RGD1310608 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 308577, 80199 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q3B756, Q9BT04 | |
| FUZ | |
| Recombinant protein encompassing a sequence within the center region of human FUZ. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction