Learn More
FUT9 Mouse, anti-Human, CoraLite Plus 488, Clone: 5D4G6, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | FUT9 |
|---|---|
| Clone | 5D4G6 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18074255
|
Proteintech
CL488-60230-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18095575
|
Proteintech
CL488-60230-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The FUT9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FUT9. This antibody recognizes human, pig, mouse, rat antigen. The FUT9 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| FUT9 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 10690 | |
| IgG2a | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 5D4G6 | |
| Immunofluorescence | |
| Liquid | |
| Human, Pig, Mouse, Rat | |
| Alpha (1,3) fucosyltransferase, Fuc TIX, Fucosyltransferase 9, Fucosyltransferase IX, FucT IX, FUT9 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.