missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUT8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12560-1-AP-20UL
This item is not returnable.
View return policy
Description
Catalyzes the addition of fucose in alpha 1-6 linkage to the first GlcNAc residue, next to the peptide chains in N-glycans.Specifications
| FUT8 | |
| Polyclonal | |
| Unconjugated | |
| FUT8 | |
| Fut8 | |
| FUT8 Fusion Protein Ag3280 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6EV76, Q9BYC5, Q9WTS2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2530, 432392, 53618 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction