missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUT7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18197-1-AP-20UL
This item is not returnable.
View return policy
Description
May catalyze alpha-1,3 glycosidic linkages involved in the expression of sialyl Lewis X antigens.Specifications
| FUT7 | |
| Polyclonal | |
| Unconjugated | |
| FUT7 | |
| Alpha (1,3) fucosyltransferase, Fuc TVII, Fucosyltransferase 7, Fucosyltransferase VII, FucT VII, FUT7, Selectin ligand synthase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14347, 2529, 296564 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q11130, Q11131 | |
| Fut7 | |
| FUT7 Fusion Protein Ag12475 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction