missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FUT6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524850
This item is not returnable.
View return policy
Description
FUT6 Polyclonal Antibody for Western Blot, IHC (P), Flow
FUT6 is a Golgi stack membrane protein that is involved in the creation of sialyl-Lewis X, an E-selectin ligand.
Specifications
| FUT6 | |
| Polyclonal | |
| Unconjugated | |
| FUT6 | |
| alpha-(1,3)-fucosyltransferase 6; FCT3A; FT1A; Fucosyltransferase 6; fucosyltransferase 6 (alpha (1,3) fucosyltransferase); fucosyltransferase VI; Fuc-TVI; FucT-VI; FUT6; Galactoside 3-L-fucosyltransferase | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 2528 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P51993 | |
| FUT6 | |
| KLH conjugated synthetic peptide between 102-130 amino acids from the central region of human FUT6. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction