missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUT4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22141-1-AP-150UL
This item is not returnable.
View return policy
Description
22141-1-AP detects the band around 45 kDa and glycosylated isoform 63 kDa protein in SDS-PAGE.
FUT4 (Fucosyltransferase 4) is a Protein Coding gene. Diseases associated with FUT4 include Colon Adenocarcinoma and Liver Lymphoma. Among its related pathways are Glycosphingolipid biosynthesis - lacto and neolacto series and Mannose type O-glycan biosynthesis. Gene Ontology (GO) annotations related to this gene include fucosyltransferase activity and alpha-(1->3)-fucosyltransferase activity. The product of this gene transfers fucose to N-acetyllactosamine polysaccharides to generate fucosylated carbohydrate structures. It catalyzes the synthesis of the non-sialylated antigen, Lewis x (CD15).Specifications
| FUT4 | |
| Polyclonal | |
| Unconjugated | |
| FUT4 | |
| Alpha (1,3) fucosyltransferase, ELFT, FCT3A, FUC TIV, Fucosyltransferase 4, Fucosyltransferase IV, FucT IV, FUT4, FUTIV | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2526 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.46 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P22083 | |
| FUT4 | |
| FUT4 Fusion Protein Ag17565 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction