missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUT4 Monoclonal Antibody (476CT26.8.4)

FUT4 Monoclonal Antibody (476CT26.8.4)
Brand: Thermo Scientific MA537568
This item is not returnable.
View return policy
Description
FUT4 Monoclonal Antibody for Western Blot
Specifications
| FUT4 | |
| Monoclonal | |
| Unconjugated | |
| FUT4 | |
| 3-fucosyl-N-acetyl-lactosamine epitope; AI451562; alpha (1,3) fucosyltransferase, myeloid specific; alpha (1,3) fucosyltransferase, myeloid-specific; alpha 1,3-fucosyltransferase; alpha 1,3-fucosyltransferase Fuc-T (similar to mouse Fut4); alpha 13-fucosyltransferase Fuc-T (similar to mouse Fut4); alpha-(1,3)-fucosyltransferase 4; CD15; ELAM ligand fucosyltransferase; ELAM-1 ligand fucosyltransferase; ELFT; FAL; FCT3A; fucosyltransferase 4; fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific); fucosyltransferase IV; Fuct; FUC-TIV; FucT-IV; FUT4; FUTIV; Galactoside 3-L-fucosyltransferase; Lewis x; LeX; Ssea1; SSEA-1; stage-specific embryonic antigen 1 | |
| Mouse | |
| Euglobulin Precipitation | |
| RUO | |
| 2526 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 476CT26.8.4 | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P22083 | |
| FUT4 | |
| KLH conjugated synthetic peptide between 425-454 amino acids from human FUT4. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction