missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FUT3 Monoclonal Antibody (1F8G5C7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548463
This item is not returnable.
View return policy
Description
FUT3 Monoclonal Antibody for Western Blot, Flow, ELISA
FUT3 comprises a set of fucosylated glycosphingolipids that are synthesized by exocrine epithelial cells and circulate in body fluids. The glycosphingolipids function in embryogenesis, tissue differentiation, tumor metastasis, inflammation, and bacterial adhesion. They are secondarily absorbed to red blood cells giving rise to their Lewis phenotype. This protein is a member of the fucosyltransferase family, which catalyzes the addition of fucose to precursor polysaccharides in the last step of Lewis antigen biosynthesis. It encodes an enzyme with alpha(1,3)-fucosyltransferase and alpha(1,4)-fucosyltransferase activities.
Specifications
| FUT3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P21217 | |
| FUT3 | |
| Purified recombinant fragment of human CD174 (AA: 199-361) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 1F8G5C7 | |
| Unconjugated | |
| FUT3 | |
| 4-fucosyltransferase; alpha-(1,3/1,4)-fucosyltransferase; blood group Lewis alpha-4-fucosyltransferase; CD174; FT3B; Fucosyltransferase 3; fucosyltransferase 3 (galactoside 3(4)-L-fucosyltransferase, Lewis blood group); fucosyltransferase 3 (Lewis blood group); fucosyltransferase III; FucT-III; Fuc-TV; FucT-V; FUT3; Galactoside 3(4)-L-fucosyltransferase; LE; Les; Lewis FT; Lewis Y | |
| Mouse | |
| Protein G | |
| RUO | |
| 2525 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction