missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FUT3/FUT5 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547569
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
FUT3 comprises a set of fucosylated glycosphingolipids that are synthesized by exocrine epithelial cells and circulate in body fluids. The glycosphingolipids function in embryogenesis, tissue differentiation, tumor metastasis, inflammation, and bacterial adhesion. They are secondarily absorbed to red blood cells giving rise to their Lewis phenotype. This protein is a member of the fucosyltransferase family, which catalyzes the addition of fucose to precursor polysaccharides in the last step of Lewis antigen biosynthesis. It encodes an enzyme with alpha(1,3)-fucosyltransferase and alpha(1,4)-fucosyltransferase activities.
Specifications
| FUT3/FUT5 | |
| Polyclonal | |
| Unconjugated | |
| FUT3 | |
| 4-fucosyltransferase; alpha (1,3) fucosyltransferase; alpha-(1,3)-fucosyltransferase 5; alpha-(1,3/1,4)-fucosyltransferase; blood group Lewis alpha-4-fucosyltransferase; CD174; FT3B; Fucosyltransferase 3; fucosyltransferase 3 (galactoside 3(4)-L-fucosyltransferase, Lewis blood group); fucosyltransferase 3 (Lewis blood group); fucosyltransferase 5; fucosyltransferase 5 (alpha (1,3) fucosyltransferase); fucosyltransferase III; Fucosyltransferase V; FucT-III; FUC-TV; fucT-V; FUT3; FUT5; Galactoside 3(4)-L-fucosyltransferase; galactoside 3-L-fucosyltransferase; LE; Les; Lewis FT; Lewis Y | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 2525, 2527 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P21217, Q11128 | |
| FUT3, FUT5 | |
| Mouse myeloma cell line NS0-derived recombinant human FUT5 Arg35-Thr374. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction