missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FUT2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA537863
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
The protein encoded by this gene is a Golgi stack membrane protein that is involved in the creation of a precursor of the H antigen, which is required for the final step in the soluble A and B antigen synthesis pathway. This gene is one of two encoding the galactoside 2-L-fucosyltransferase enzyme. Two transcript variants encoding the same protein have been found for this gene.
Specifications
| FUT2 | |
| Polyclonal | |
| Unconjugated | |
| FUT2 | |
| 12FT 2; 12FT2; alpha (1,2) fucosyltransferase; alpha(1,2)FT 2; alpha(1,2)FT2; B12QTL1; Fucosyltransferase 2; fucosyltransferase 2 (secretor status included); FUT2; Galactoside 2-alpha-L-fucosyltransferase 2; GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 2; Se; SE2; SEC2; Secretor blood group alpha-2-fucosyltransferase; secretor factor; Secretory blood group protein 2; sej | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 2524 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q10981 | |
| FUT2 | |
| Peptide with sequence C-HVQQRLAKIQAMWE, from the internal region near the N-terminus of FUT2 (aa 27-40). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction