missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FUT10 Polyclonal antibody specifically detects FUT10 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | FUT10 |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 646 |
| Dilution | ELISA |
| Gene Alias | alpha 1,3-fucosyl transferase, alpha-(1,3)-fucosyltransferase 10, EC 2.4.1, EC 2.4.1.-, Fucosyltransferase 10, fucosyltransferase 10 (alpha (1,3) fucosyltransferase), Fucosyltransferase X, Fuc-TX, FucT-X, Galactoside 3-L-fucosyltransferase 10, MGC11141 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human FUT10 (Uniprot#: Q6P4F1-1; Leu 32-Asp 479) |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Content And Storage | Store at 4C in the dark. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?