missing translation for 'onlineSavingsMsg'
Learn More

FUT10 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18390264 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390264 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390264 Supplier Novus Biologicals Supplier No. NBP299776AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FUT10 Polyclonal specifically detects FUT10 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUT10
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias alpha 1,3-fucosyl transferase, alpha-(1,3)-fucosyltransferase 10, EC 2.4.1, EC 2.4.1.-, Fucosyltransferase 10, fucosyltransferase 10 (alpha (1,3) fucosyltransferase), Fucosyltransferase X, Fuc-TX, FucT-X, Galactoside 3-L-fucosyltransferase 10, MGC11141
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant FUT10 (Q6P4F1-1; Leu 32-Asp 479).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 84750
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.