missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUT10 Antibody (003), Alexa Fluor™ 350 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89889AF350
This item is not returnable.
View return policy
Description
FUT10 Monoclonal antibody specifically detects FUT10 in Human samples. It is validated for ELISA
Specifications
| FUT10 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| alpha 1,3-fucosyl transferase, alpha-(1,3)-fucosyltransferase 10, EC 2.4.1, EC 2.4.1.-, Fucosyltransferase 10, fucosyltransferase 10 (alpha (1,3) fucosyltransferase), Fucosyltransferase X, Fuc-TX, FucT-X, Galactoside 3-L-fucosyltransferase 10, MGC11141 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human FUT10 (Q6P4F1-1; Leu 32-Asp 479). | |
| 0.1 mL | |
| Neuroscience | |
| 84750 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 003 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction