missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fusion glycoprotein F0 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109300
This item is not returnable.
View return policy
Description
Positive Control: Fusion glycoprotein F0 transfected HEK293 cell.
During virus entry, induces fusion of viral and cellular membranes leading to delivery of the nucleocapsid into the cytoplasm. The fusogenic activity is inactive untill entry into host cell endosome, where a furin-like protease cleaves off a small peptide between F1 and F2. Interacts directly with heparan sulfate and may participate in virus attachment. Furthermore, the F2 subunit was identifed as the major determinant of RSV host cell specificity. Later in infection, proteins F expressed at the plasma membrane of infected cells can mediate fusion with adjacent cells to form syncytia, a cytopathic effect that could lead to tissue necrosis. The fusion protein is also able to trigger p53-dependent apoptosis.
Specifications
| Fusion glycoprotein F0 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Synthetic peptide within newcastle disease virus Fusion glycoprotein F0 aa 504-553. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction