missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fusidic Acid Hemihydrate, Mikromol™
Description
Fusidic Acid Hemihydrate
Specifications
Specifications
| Chemical Name or Material | Fusidic acid hemihydrate |
| CAS | 172343-30-5 |
| Analyte or Component Names | Fusidic acid hemihydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2 C31 H48 O6 . H2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Fenoprofen 1,3-Butylene Glycol Esters (Mixture of Regio- and Stereoisomers), ent-(17Z)-16alpha-(Acetyloxy)-3beta,11beta-dihydroxy-4beta,8,14-trimethyl-18-nor-5beta,10alpha-cholesta-17(20),24-dien-21-oic acid hemihydrate, 29-Nordammara-17(20),24-dien-21-oic acid, 16-(acetyloxy)-3,11-dihydroxy-, hydrate (2:1), (3alpha,4alpha,8alpha,9beta,11alpha,13alpha,14beta,16beta,17Z)-, Fusidic acid hemihydrate |
| InChI Formula | InChI=1S/2C31H48O6.H2O/c2*1-17(2)9-8-10-20(28(35)36)26-22-15-24(34)27-29(5)13-12-23(33)18(3)21(29)11-14-30(27,6)31(22,7)16-25(26)37-19(4)32;/h2*9,18,21-25,27,33-34H,8,10-16H2,1-7H3,(H,35,36);1H2/b2*26-20-;/t2*18-,21-,22-,23+,24+,25-,27-,29-,30-,31-;/m00./s1 |
| SMILES | O.C[C@@H]1[C@H](O)CC[C@@]2(C)[C@H]1CC[C@@]3(C)[C@H]2[C@H](O)C[C@H]4\C(=C(/CCC=C(C)C)\C(=O)O)\[C@H](C[C@]34C)OC(=O)C.C[C@@H]5[C@H](O)CC[C@@]6(C)[C@H]5CC[C@@]7(C)[C@H]6[C@H](O)C[C@H]8\C(=C(/CCC=C(C)C)\C(=O)O)\[C@H](C[C@]78C)OC(=O)C |
| IUPAC Name | (2Z)-2-[(3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetyloxy-3,11-dihydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ylidene]-6-methylhept-5-enoic acid;hydrate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?