missing translation for 'onlineSavingsMsg'
Learn More

Fusidic acid, 98%, Thermo Scientific Chemicals

Product Code. 15552333 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15552333 5 g 5g
15542333 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15552333 Supplier Thermo Scientific Acros Supplier No. 458460050
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6990-06-3

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Molecular Formula C31H48O6
Quantity 5 g
Synonym methoxyphenylacetic acid, methoxy phenyl acetic acid, dl-alpha-methoxyphenylacetic acid, acetic acid, methoxyphenyl, alpha-methoxyphenylacetic acid, o-methyl-dl-mandelic acid, benzeneacetic acid, .alpha.-methoxy, 2-methoxy-2-phenyl-acetic acid, methyloxy phenyl acetic acid, .alpha.-methoxyphenylacetic acid
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol,chloroform
InChI Key DIWVBIXQCNRCFE-UHFFFAOYNA-N
SMILES COC(C1=CC=CC=C1)C(=O)O
IUPAC Name 2-methoxy-2-phenylacetic acid
Molecular Weight (g/mol) 516.71
PubChem CID 107202
Formula Weight 516.71
Percent Purity 98%
Chemical Name or Material Fusidic acid
Show More Show Less
compliance-icons
Product Identifier
  • Fusidic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.