missing translation for 'onlineSavingsMsg'
Learn More

Fusaric acid, 99%

Product Code. 10197693 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10197693 250 mg Glass bottle 250mg
10411631 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10197693 Supplier Thermo Scientific Acros Supplier No. 198962500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 536-69-6 | C10H13NO2 | 179.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-69-6
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00006298
InChI Key DGMPVYSXXIOGJY-UHFFFAOYSA-N
Synonym fusaric acid, 5-butylpicolinic acid, fusarinic acid, 2-pyridinecarboxylic acid, 5-butyl, 5-butyl-2-pyridinecarboxylic acid, picolinic acid, 5-butyl, 5-n-butylpyridine-2-carboxylic acid, unii-jwj963070n, 5-butylpyridine-3-carboxylic acid, 5-butyl-pyridine-2-carboxylic acid
PubChem CID 3442
ChEBI CHEBI:5199
IUPAC Name 5-butylpyridine-2-carboxylic acid
SMILES CCCCC1=CN=C(C=C1)C(=O)O

Specifications

Melting Point 97°C to 101°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 4343
Formula Weight 179.22
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Fusaric acid
Show More Show Less
compliance-icons
Product Identifier
  • Fusaric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.