missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FUS2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15796711 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15796711 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15796711 Supplier Invitrogen™ Supplier No. PA558844

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PSLAELTLEP VHRRPELLDA CADLINDQWP RSRTSRLHSL GQSSDAFPLC LMLLSPHPTL EAAPVVVGHA RLSRVLN Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.

This gene encodes a member of the N-acetyltransferase family. N-acetyltransferases modify proteins by transferring acetyl groups from acetyl CoA to the N-termini of protein substrates. The encoded protein is a cytoplasmic N-acetyltransferase with a substrate specificity for proteins with an N-terminal methionine. This gene is located in the tumor suppressor gene region on chromosome 3p21.3 and the encoded protein may play a role in cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed. This gene overlaps and is on the same strand as hyaluronoglucosaminidase 3, and some transcripts of each gene share a portion of the first exon.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUS2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NAA80
Gene Accession No. Q93015, Q9R123
Gene Alias AI225910; Fus2; FUS-2; fusion 2; HsNAAA80; LOC100910872; NAA80; N-acetyltransferase 6; N-acetyltransferase 6 (GCN5-related); N-acetyltransferase 6-like; N-alpha-acetyltransferase 80; NAT6; Protein fus-2; protein fusion-2; Protein fusion-2 homolog
Gene Symbols NAA80
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FUS2. Recombinant protein control fragment (Product #RP-96495).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100910872, 24142, 56441
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.