missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUS2 Antibody (2F4), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-41157FR
This item is not returnable.
View return policy
Description
FUS2 Monoclonal specifically detects FUS2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| FUS2 | |
| Monoclonal | |
| DyLight 680 | |
| NP_036323 | |
| NAA80 | |
| Recombinant FUS2 (1-308aa) purified from E. coli | |
| 0.1 mL | |
| Cancer, Proteases & Other Enzymes | |
| 24142 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 2F4 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| FUS-2, FUS2EC 2.3.1.-, N-acetyltransferase 6, N-acetyltransferase 6 (GCN5-related), Protein fus-2, Protein fusion-2 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human FUS2 protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction