missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FUS Monoclonal Antibody (1FU-1D2)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA3089

Product Code. 15865873

  • £440.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA3-089 detects AID from human samples MA3-089 has been successfully used in Western blot. The MA3-089 immunogen is the synthetic peptide coupled to ovalalbumin, aa 266 to 283. MA3-089 detects FUS/TLS (~70kDa) in human samples.

This gene encodes a multifunctional protein component of the heterogeneous nuclear ribonucleoprotein (hnRNP) complex. The hnRNP complex is involved in pre-mRNA splicing and the export of fully processed mRNA to the cytoplasm. This protein belongs to the FET family of RNA-binding proteins which have been implicated in cellular processes that include regulation of gene expression, maintenance of genomic integrity and mRNA/microRNA processing. Alternative splicing results in multiple transcript variants. Defects in this gene result in amyotrophic lateral sclerosis type 6.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FUS
Monoclonal
Conc. Not Determined
Ascites with 0.05% sodium azide
P35637
FUS
Synthetic peptide (GPRDQGSRHDSEQDNSDC) coupled to ovalbumin. From aa 266 to aa 283 of FUS.
RUO
2521
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot, Immunocytochemistry
1FU-1D2
Unconjugated
FUS
16) in malignant liposarcoma); 16) malignant liposarcoma; 75 kDa DNA-pairing protein; ALS6; D430004D17Rik; D930039C12Rik; ETM4; FUS; FUS RNA binding protein; Fus1; fused in sarcoma; fused in sarcoma RNA binding protein; fusion (involved in t(12; fusion gene in myxoid liposarcoma; fusion, derived from t(12; fusion, derived from t(12;16) malignant liposarcoma; fus-like protein; heterogeneous nuclear ribonucleoprotein P2; hnRNP P2; HNRNPP2; oncogene FUS; Oncogene TLS; pigpen protein; POMP75; Protein pigpen; RNA-binding protein FUS; TLS; translocated in liposarcoma; translocated in liposarcoma protein
Mouse
50 μL
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.