missing translation for 'onlineSavingsMsg'
Learn More

Fursultiamine, MedChemExpress

Product Code. 16433816
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16433816

Brand: Medchem Express HYB2082

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fursultiamine is a vitamin B1 derivative, has anti-nociceptive and antineoplastic activity. Fursultiamine can be used for vitamin B1 deficiency, osteoarthritis (OA) and cancer research.

Fursultiamine is a vitamin B1 derivative, has anti-nociceptive and antineoplastic activity. Fursultiamine can be used for vitamin B1 deficiency, osteoarthritis (OA) and cancer research.

Specifications

Chemical Name or Material Fursultiamine
CAS 804-30-8
Color White
Purity Grade Notes Research
Molecular Formula C17H26N4O3S2
Quantity 25 mg
Solubility Information DMSO : 106.7 mg/mL (267.73 mM; Need ultrasonic and warming)
SMILES O=CN(CC1=CN=C(C)N=C1N)/C(C)=C(SSCC2OCCC2)\CCO
Molecular Weight (g/mol) 398.54
Formula Weight 398.54
Percent Purity 99.86%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.