missing translation for 'onlineSavingsMsg'
Learn More

Fursultiamine, MedChemExpress

Product Code. 16498728
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498728 10 mg 10mg
16478836 100 mg 100mg
16433816 25 mg 25mg
16424647 50 mg 50mg
16421148 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16498728 Supplier Medchem Express Supplier No. HYB2082
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fursultiamine is a vitamin B1 derivative, has anti-nociceptive and antineoplastic activity. Fursultiamine can be used for vitamin B1 deficiency, osteoarthritis (OA) and cancer research.

Fursultiamine is a vitamin B1 derivative, has anti-nociceptive and antineoplastic activity. Fursultiamine can be used for vitamin B1 deficiency, osteoarthritis (OA) and cancer research.

Specifications

Chemical Name or Material Fursultiamine
CAS 804-30-8
Color White
Purity Grade Notes Research
Molecular Formula C17H26N4O3S2
Quantity 10 mg
Solubility Information DMSO : 106.7 mg/mL (267.73 mM; Need ultrasonic and warming)
SMILES O=CN(CC1=CN=C(C)N=C1N)/C(C)=C(SSCC2OCCC2)\CCO
Molecular Weight (g/mol) 398.54
Formula Weight 398.54
Percent Purity 99.86%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.