missing translation for 'onlineSavingsMsg'
Learn More

Furoyl Chloride, TRC

Product Code. 30400990
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400990 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400990 Supplier Toronto Research Chemicals Supplier No. TRCF86520025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Furoyl Chloride
CAS 527-69-5
Molecular Formula C5H3ClO2
Quantity 25 g
Synonym 2-Furancarbonyl Chloride, 2-Furoyl Chloride, 2-(Chlorocarbonyl)furan, 2-(Chloroformyl)furan, 2-Furancarboxylic Acid Chloride, 2-Furanoic Acid Chloride, 2-Furanoyl Chloride, 2-Furanylcarbonyl Chloride, 2-Furoic Acid Chloride, 2-Furoic Chloride, 2-Furylcarbonyl Chloride, Furan-2-carbonyl Chloride, Furan-2-carboxylic Chloride, Pyromucyl Chloride, alpha-Furoic Chloride
InChI Formula InChI=1S/C5H3ClO2/c6-5(7)4-2-1-3-8-4/h1-3H
SMILES ClC(=O)c1occc1
Molecular Weight (g/mol) 130.53
Formula Weight 129.9822
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.