missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Furoyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | Furoyl Chloride |
| CAS | 527-69-5 |
| Molecular Formula | C5H3ClO2 |
| Quantity | 250 g |
| Synonym | 2-Furancarbonyl Chloride, 2-Furoyl Chloride, 2-(Chlorocarbonyl)furan, 2-(Chloroformyl)furan, 2-Furancarboxylic Acid Chloride, 2-Furanoic Acid Chloride, 2-Furanoyl Chloride, 2-Furanylcarbonyl Chloride, 2-Furoic Acid Chloride, 2-Furoic Chloride, 2-Furylcarbonyl Chloride, Furan-2-carbonyl Chloride, Furan-2-carboxylic Chloride, Pyromucyl Chloride, alpha-Furoic Chloride |
| InChI Formula | InChI=1S/C5H3ClO2/c6-5(7)4-2-1-3-8-4/h1-3H |
| SMILES | ClC(=O)c1occc1 |
| Molecular Weight (g/mol) | 130.53 |
| Formula Weight | 129.9822 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Vous avez repéré une opportunité d'amélioration ?