missing translation for 'onlineSavingsMsg'
Learn More

Furosemide, TRC

Product Code. 18053440
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18053440 10 g 10g
18071930 1 g 1g
18012140 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18053440 Supplier Toronto Research Chemicals Supplier No. TRCF86500010G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-31-9
Molecular Formula C12 H11 Cl N2 O5 S
Molecular Weight (g/mol) 330.74
Synonym Benzoic acid, 5-(aminosulfonyl)-4-chloro-2-[(2-furanylmethyl)amino]-, Anthranilic acid, 4-chloro-N-furfuryl-5-sulfamoyl-, 2-Furfurylamino-4-chloro-5-sulfamoylbenzoic acid, 4-Chloro-N-(2-furylmethyl)-5-sulfamoylanthranilic acid, 4-Chloro-N-furfuryl-5-sulfamoylanthranilic acid, 5-(Aminosulfamyl)-4-chloro-2-[(2-furanylmethyl)amino]benzoic acid, 5-(Aminosulfonyl)-4-chloro-2-[(2-furanylmethyl)amino]benzoic acid, Aisemide, Aldic, Aluzine, Anfuramaide, Apo-Frusemide, Apo-Furosemide, Aquamide, Aquarid, Aquasin, Arasemide, Beronald, Bioretic, Cetasix, Desal, Desdemin, Dirine, Disal, Discoid, Disemide, Diural, Diuresal, Diuretic salt, Diurin, Diurolasa, Diusemide, Diusil, Dranex, Dryptal, Durafurid, Errolon, Eutensin, Fluidrol, Franyl, Frumex, Frumide, Frusedan, Frusema, Frusemide, Frusemin, Frusenex, Frusetic, Frusid, Fulsix, Fuluvamide, Furanthril, Furanthryl, Furantril, Furanturil, Furesis, Furetic, Furex, Furix, Furmid, Furo-Basan, Furo-Puren, Furocot, Furodiurol, Furomen, Furomex, Furomide M.D., Furorese, Furosan, Furosedon, Furosemid, Furosemide, Furosix, Furoter, Furovite, Fusid, Golan, Hissuflux, Hydrex, Hydro, Hydro-rapid, Impugan, Katlex, Kofuzon, Kutrix, LB 502, Lasemid, Lasex, Lasiletten, Lasilix, Lasix, Lasix Retard, Laxur, Liside, Lowpston, Lowpstron, Macasirool, Marsemide, Mirfat, NSC 269420, Naclex, Nadis, Nelsix, Nephron, Nicorol, Odemase, Odemex, Oedemex, Prefemin, Profemin, Promedes, Promide, Radisemide, Radonna, Retep, Rosemide, Rusyde, Salinex, Salix, Salurid, Seguril, Selectofur, Transit, Trofurit, Uremide, Urenil, Uresix, Urex, Urex M, Urian, Uridon, Yidoli, Zafurida
IUPAC Name 4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid
SMILES NS(=O)(=O)c1cc(C(=O)O)c(NCc2occc2)cc1Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H11ClN2O5S/c13-9-5-10(15-6-7-2-1-3-20-7)8(12(16)17)4-11(9)21(14,18)19/h1-5,15H,6H2,(H,16,17)(H2,14,18,19)
Formula Weight 330.0077
Percent Purity >95%
Chemical Name or Material Furosemide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.