missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Furosemide sodium, MedChemExpress
Description
Furosemide sodium is a potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter, NKCC1 and NKCC2. Furosemide sodium is also a GABAA receptors antagonist and displays 100-fold selectivity for α6-containing receptors than α1-containing receptors. Furosemide sodium acts as a loop diuretic and used for the study of congestive heart failure, hypertension and edema.
Specifications
Specifications
| Chemical Name or Material | Furosemide sodium |
| CAS | 41733-55-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C12H10ClN2NaO5S |
| Quantity | 1 g |
| Solubility Information | DMSO : ≥ 150 mg/mL (425.25 mM) ∣H2O : 100 mg/mL (283.50 mM; Need ultrasonic) |
| SMILES | O=C(C1=CC(S(=O)(N)=O)=C(C=C1NCC2=CC=CO2)Cl)O[Na] |
| Molecular Weight (g/mol) | 352.73 |
| Formula Weight | 352.73 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?