missing translation for 'onlineSavingsMsg'
Learn More

Furosemide, MedChemExpress

Product Code. 16412059
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16412059 1 g 1g
16400788 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16412059 Supplier Medchem Express Supplier No. HYB0135
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Furosemide is a potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter, NKCC1 and NKCC2. Furosemide is also a GABAA receptors antagonist and displays 100-fold selectivity for α6-containing receptors than α1-containing receptors. Furosemide acts as a loop diuretic and used for the study of congestive heart failure, hypertension and edema.

Furosemide is a potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter, NKCC1 and NKCC2. Furosemide is also a GABAA receptors antagonist and displays 100-fold selectivity for α6-containing receptors than α1-containing receptors. Furosemide acts as a loop diuretic and used for the study of congestive heart failure, hypertension and edema.

Specifications

Chemical Name or Material Furosemide
CAS 54-31-9
Color White
Purity Grade Notes Research
Molecular Formula C12H11ClN2O5S
Quantity 1 g
Solubility Information DMSO : ≥ 100 mg/mL (302.35 mM) ∣H2O : 0.1 mg/mL (0.30 mM; Need ultrasonic)
SMILES O=C(O)C1=CC(S(=O)(N)=O)=C(Cl)C=C1NCC2=CC=CO2
Molecular Weight (g/mol) 330.74
Formula Weight 330.74
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.