missing translation for 'onlineSavingsMsg'
Learn More

Furosemide, LoGiCal

Product Code. 30461656
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461656 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461656 Supplier LoGiCal Supplier No. LGCFOR0014.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Furosemide
CAS 54-31-9
Analyte or Component Names Furosemide
Shipping Condition Room Temperature
Molecular Formula C12 H11 Cl N2 O5 S
Certifications/Compliance ISO 9001
Synonym Benzoic acid, 5-(aminosulfonyl)-4-chloro-2-[(2-furanylmethyl)amino]-, Anthranilic acid, 4-chloro-N-furfuryl-5-sulfamoyl-, 2-Furfurylamino-4-chloro-5-sulfamoylbenzoic acid, 4-Chloro-N-(2-furylmethyl)-5-sulfamoylanthranilic acid, 4-Chloro-N-furfuryl-5-sulfamoylanthranilic acid, 5-(Aminosulfamyl)-4-chloro-2-[(2-furanylmethyl)amino]benzoic acid, 5-(Aminosulfonyl)-4-chloro-2-[(2-furanylmethyl)amino]benzoic acid, Aisemide, Aldic, Aluzine, Anfuramaide, Apo-Frusemide, Apo-Furosemide, Aquamide, Aquarid, Aquasin, Arasemide, Beronald, Bioretic, Cetasix, Desal, Desdemin, Dirine, Disal, Discoid, Disemide, Diural, Diuresal, Diuretic salt, Diurin, Diurolasa, Diusemide, Diusil, Dranex, Dryptal, Durafurid, Errolon, Eutensin, Fluidrol, Franyl, Frumex, Frumide, Frusedan, Frusema, Frusemide, Frusemin, Frusenex, Frusetic, Frusid, Fulsix, Fuluvamide, Furanthril, Furanthryl, Furantril, Furanturil, Furesis, Furetic, Furex, Furix, Furmid, Furo-Basan, Furo-Puren, Furocot, Furodiurol, Furomen, Furomex, Furomide M.D., Furorese, Furosan, Furosedon, Furosemid, Furosemide, Furosix, Furoter, Furovite, Fusid, Golan, Hissuflux, Hydrex, Hydro, Hydro-rapid, Impugan, Katlex, Kofuzon, Kutrix, LB 502, Lasemid, Lasex, Lasiletten, Lasilix, Lasix, Lasix Retard, Laxur, Liside, Lowpston, Lowpstron, Macasirool, Marsemide, Mirfat, NSC 269420, Naclex, Nadis, Nelsix, Nephron, Nicorol, Odemase, Odemex, Oedemex, Prefemin, Profemin, Promedes, Promide, Radisemide, Radonna, Retep, Rosemide, Rusyde, Salinex, Salix, Salurid, Seguril, Selectofur, Transit, Trofurit, Uremide, Urenil, Uresix, Urex, Urex M, Urian, Uridon, Yidoli, Zafurida
InChI Formula InChI=1S/C12H11ClN2O5S/c13-9-5-10(15-6-7-2-1-3-20-7)8(12(16)17)4-11(9)21(14,18)19/h1-5,15H,6H2,(H,16,17)(H2,14,18,19)
Solution Type Neat
SMILES NS(=O)(=O)c1cc(C(=O)O)c(NCc2occc2)cc1Cl
IUPAC Name 4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 330.74
Quantity 10 mg
Formula Weight 330.0077 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.