missing translation for 'onlineSavingsMsg'
Learn More

Furo[3,2-b]pyridine, 95%

Product Code. 15475757
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475757 5 g 5g
15485757 250 mg 250mg
15465757 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15475757 Supplier Thermo Scientific Alfa Aesar Supplier No. H32966.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 272-62-8 | C7H5NO | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 272-62-8
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.12
MDL Number MFCD11870744
InChI Key YRTCKZIKGWZNCU-UHFFFAOYSA-N
Synonym furo 3,2-b pyridine, 4-azabenzo b furan, furopyridine, furo 3,2-b pyridin, 4-aza-1-benzofuran
PubChem CID 12210217
IUPAC Name furo[3,2-b]pyridine
SMILES O1C=CC2=NC=CC=C12

Specifications

Boiling Point 79°C to 80°C (12 mmHg)
Refractive Index 1.5753
Quantity 5 g
Formula Weight 119.12
Percent Purity 98%
Chemical Name or Material Furo[3,2-b]pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.