missing translation for 'onlineSavingsMsg'
Learn More

Carbolite™ Furnace with Type R Temperature Sensor

Brand:  Carbolite™ RHF1515-230SN+&02-301


Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12183290

  • £13635.00 / Each
Estimated Shipment: 20-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Compliance to BSEN 61010
15 L
Furnace
305 x 220 x 220 mm
220 mm
220 mm
305 mm
230 V
50/60 Hz
301 PID Controller
1500°C
For use in quality control, heat treatment and research of ceramic materials.
690 mm
810 mm
780 mm
230 V 50/60 Hz 10000 w
10000 w
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers