missing translation for 'onlineSavingsMsg'
Learn More

Furfurylamine, 99%, Thermo Scientific Chemicals

Catalog Number p-7027313
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
617-89-0
C5H7NO
97.117
MFCD00003258
DDRPCXLAQZKBJP-UHFFFAOYSA-N
furfurylamine, 2-furfurylamine, 2-furanmethylamine, 2-aminomethylfuran, 2-aminomethyl furan, 2-furanmethanamine, 2-furylmethylamine, 2-furylmethyl amine, usaf q-1, methylamine, 1-2-furyl
3438
furan-2-ylmethanamine
C1=COC(=C1)CN
This item is not returnable. View return policy
617-89-0
C5H7NO
97.117
MFCD00003258
DDRPCXLAQZKBJP-UHFFFAOYSA-N
furfurylamine, 2-furfurylamine, 2-furanmethylamine, 2-aminomethylfuran, 2-aminomethyl furan, 2-furanmethanamine, 2-furylmethylamine, 2-furylmethyl amine, usaf q-1, methylamine, 1-2-furyl
3438
furan-2-ylmethanamine
C1=COC(=C1)CN

Furfurylamine is used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics, antihypertensive, and antiseptic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furfurylamine is used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics, antihypertensive, and antiseptic agents.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are strong oxidizing agents, air, acids.
TRUSTED_SUSTAINABILITY
Melting Point -70°C
Density 1.05
pH 11.6
Boiling Point 145°C to 146°C
Flash Point 37°C (99°F)
Odor Amine-like
Refractive Index 1.49
Quantity 100 g
UN Number UN2526
Beilstein 1614
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 97.12
Percent Purity 99%
Chemical Name or Material Furfurylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'