missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl Mercaptan, TRC

Product Code. 30407037
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407037 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30407037 Supplier Toronto Research Chemicals Supplier No. TRCF8643005G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Furan-2-ylmethanethiol
CAS 98-02-2
Molecular Formula C5 H6 O S
Quantity 5 g
Synonym 2-Furanmethanethiol (6CI, 7CI, 8CI, 9CI, ACI), (2-Furanyl)methylmercaptan, 2-(Mercaptomethyl)furan, 2-Furfurylmercaptan, 2-Furfurylthiol, 2-Furylmethanethiol, 2-Furylmethyl mercaptan, 2-Furylmethylthiol, Furan-2-ylmethanethiol, Furfuryl mercaptan, Furfuryl thiol, Furfurylthiol, NSC 41142
InChI Formula InChI=1S/C5H6OS/c7-4-5-2-1-3-6-5/h1-3,7H,4H2
SMILES SCc1occc1
IUPAC Name furan-2-ylmethanethiol
Molecular Weight (g/mol) 114.165
Formula Weight 114.014
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.