missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl mercaptan, 98+%

Product Code. 11410487
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410487 25 g 25g
11420487 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410487 Supplier Thermo Scientific Alfa Aesar Supplier No. B22380.14
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 98-02-2 | C5H6OS | 114.162 g/mol

Furfuryl Mercaptam is a volatile flavor component of corn tortilla chips.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furfuryl Mercaptam is a volatile flavor component of corn tortilla chips.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-02-2
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.162
MDL Number MFCD00003254
InChI Key ZFFTZDQKIXPDAF-UHFFFAOYSA-N
Synonym furfuryl mercaptan, 2-furanmethanethiol, 2-furylmethanethiol, furfuryl thiol, 2-furylmethyl mercaptan, 2-furfurylmercaptan, furan-2-yl-methanethiol, 2-mercaptomethyl furan, furfurylmercaptan, 2-furfurylthiol
PubChem CID 7363
IUPAC Name furan-2-ylmethanethiol
SMILES C1=COC(=C1)CS

Specifications

Density 1.132
Boiling Point 155°C
Flash Point 45°C (113°F)
Odor Strong
Refractive Index 1.5315
Quantity 25 g
UN Number UN1228
Beilstein 383594
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 114.17
Percent Purity ≥98%
Chemical Name or Material Furfuryl mercaptan
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.