missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl isothiocyanate, 97%

Product Code. 11494007
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494007 100 g 100g
11484007 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494007 Supplier Thermo Scientific Alfa Aesar Supplier No. A19410.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4650-60-6 | C6H5NOS | 139.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4650-60-6
Molecular Formula C6H5NOS
Molecular Weight (g/mol) 139.172
MDL Number MFCD00014112
InChI Key ICXYINJACKJQQV-UHFFFAOYSA-N
Synonym furfuryl isothiocyanate, 2-isothiocyanatomethyl furan, 2-furylmethyl isothiocyanate, 2-isothiocyanatomethyl-furan, furan, 2-isothiocyanatomethyl, 2-furanylmethyl isothiocyanate, 2-furylmethanisothiocyanate, furfurylisothiocyanate, furylmethylisothiocyanate, 7-bromo-quinolin-2-ylamine
PubChem CID 78380
IUPAC Name 2-(isothiocyanatomethyl)furan
SMILES C1=COC(=C1)CN=C=S

Specifications

Density 1.39
Boiling Point 64°C to 68°C (2 mmHg)
Odor Pungent
Refractive Index 1.5655
Quantity 100 g
UN Number UN2922
Beilstein 1281561
Sensitivity Moisture sensitive
Formula Weight 139.18
Percent Purity 97%
Chemical Name or Material Furfuryl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.