missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl isothiocyanate, 97%

Product Code. 11484007
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11484007

Brand: Thermo Scientific Alfa Aesar A19410.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4650-60-6 | C6H5NOS | 139.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4650-60-6
Molecular Formula C6H5NOS
Molecular Weight (g/mol) 139.172
MDL Number MFCD00014112
InChI Key ICXYINJACKJQQV-UHFFFAOYSA-N
Synonym furfuryl isothiocyanate, 2-isothiocyanatomethyl furan, 2-furylmethyl isothiocyanate, 2-isothiocyanatomethyl-furan, furan, 2-isothiocyanatomethyl, 2-furanylmethyl isothiocyanate, 2-furylmethanisothiocyanate, furfurylisothiocyanate, furylmethylisothiocyanate, 7-bromo-quinolin-2-ylamine
PubChem CID 78380
IUPAC Name 2-(isothiocyanatomethyl)furan
SMILES C1=COC(=C1)CN=C=S

Specifications

Density 1.39
Boiling Point 64°C to 68°C (2 mmHg)
Odor Pungent
Refractive Index 1.5655
Quantity 25 g
UN Number UN2922
Beilstein 1281561
Sensitivity Moisture sensitive
Formula Weight 139.18
Percent Purity 97%
Chemical Name or Material Furfuryl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.